Product Details

SNP ID
rs117059294
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:22455292 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAATTCTGATGACACAGTAGCTACA[A/G]AGGATTAATTTTAAAAATATTAATT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C2CD5 PubMed Links

Gene Details

Gene
C2CD5
Gene Name
C2 calcium dependent domain containing 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286173.1 Intron NP_001273102.1
NM_001286174.1 Intron NP_001273103.1
NM_001286175.1 Intron NP_001273104.1
NM_001286176.1 Intron NP_001273105.1
NM_001286177.1 Intron NP_001273106.1
NM_014802.2 Intron NP_055617.1
XM_005253538.2 Intron XP_005253595.1
XM_005253539.3 Intron XP_005253596.1
XM_006719170.2 Intron XP_006719233.1
XM_017020264.1 Intron XP_016875753.1
XM_017020265.1 Intron XP_016875754.1
XM_017020266.1 Intron XP_016875755.1
XM_017020267.1 Intron XP_016875756.1
XM_017020268.1 Intron XP_016875757.1
XM_017020269.1 Intron XP_016875758.1
XM_017020270.1 Intron XP_016875759.1
XM_017020271.1 Intron XP_016875760.1
XM_017020272.1 Intron XP_016875761.1
XM_017020273.1 Intron XP_016875762.1
XM_017020274.1 Intron XP_016875763.1
XM_017020275.1 Intron XP_016875764.1
XM_017020276.1 Intron XP_016875765.1
XM_017020277.1 Intron XP_016875766.1
XM_017020278.1 Intron XP_016875767.1
XM_017020279.1 Intron XP_016875768.1

View Full Product Details