Product Details
- SNP ID
-
rs117139010
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:26970392 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTCCCCTCTTACTGCTCTTCACACG[A/G]TCTTTCCCCTGTGAGCACATGCACC
- Phenotype
-
MIM: 608858
MIM: 605181
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
FGFR1OP2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs11381588] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- FGFR1OP2
- Gene Name
- FGFR1 oncogene partner 2
There are no transcripts associated with this gene.
- Gene
- TM7SF3
- Gene Name
- transmembrane 7 superfamily member 3
View Full Product Details