Product Details

SNP ID
rs112758033
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:27028703 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAACTGAGTGAAATTATTAAGGCAT[A/G]TAATACATTAATGAACATAATATAA
Phenotype
MIM: 603800
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MED21 PubMed Links

Gene Details

Gene
MED21
Gene Name
mediator complex subunit 21
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001271811.1 733 UTR 3 NP_001258740.1
NM_004264.4 733 UTR 3 NP_004255.2

View Full Product Details