Product Details

SNP ID
rs116694079
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:132196995 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCCCTCAGCCTCTGCTGTCCTGGC[C/T]GCACATAGAGCCCTGTCCTGCTGTG
Phenotype
MIM: 606251
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GALNT9 PubMed Links

Gene Details

Gene
GALNT9
Gene Name
polypeptide N-acetylgalactosaminyltransferase 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001122636.1 1104 UTR 3 NP_001116108.1
NM_021808.3 1104 UTR 3 NP_068580.2
XM_017019371.1 1104 UTR 3 XP_016874860.1
XM_017019372.1 1104 UTR 3 XP_016874861.1
XM_017019373.1 1104 UTR 3 XP_016874862.1
XM_017019374.1 1104 UTR 3 XP_016874863.1
XM_017019375.1 1104 Intron XP_016874864.1
XM_017019376.1 1104 Intron XP_016874865.1

View Full Product Details