Product Details
- SNP ID
-
rs117969623
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:113195640 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCCTTCTCCTTGTCCTTTGCTTTGC[C/T]CTTGCCCCGCTTCTTCTTGGATCTG
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
IQCD
PubMed Links
Gene Details
- Gene
- IQCD
- Gene Name
- IQ motif containing D
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_138451.2 |
946 |
Missense Mutation |
AGC,GGC |
S327G |
NP_612460.1 |
XM_005253832.4 |
946 |
Missense Mutation |
AGC,GGC |
S429G |
XP_005253889.1 |
XM_005253833.4 |
946 |
Missense Mutation |
AGC,GGC |
S429G |
XP_005253890.1 |
XM_005253834.4 |
946 |
Missense Mutation |
AGC,GGC |
S248G |
XP_005253891.1 |
XM_011537862.2 |
946 |
Missense Mutation |
AGC,GGC |
S429G |
XP_011536164.1 |
XM_011537863.2 |
946 |
Missense Mutation |
AGC,GGC |
S429G |
XP_011536165.1 |
XM_011537864.1 |
946 |
Missense Mutation |
AGC,GGC |
S351G |
XP_011536166.1 |
XM_011537865.2 |
946 |
Missense Mutation |
AGC,GGC |
S170G |
XP_011536167.1 |
XM_011537866.2 |
946 |
Missense Mutation |
AGC,GGC |
S89G |
XP_011536168.1 |
XM_017018771.1 |
946 |
Missense Mutation |
AGC,GGC |
S146G |
XP_016874260.1 |
- Gene
- RITA1
- Gene Name
- RBPJ interacting and tubulin associated 1
There are no transcripts associated with this gene.
View Full Product Details