Product Details

SNP ID
rs113219038
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:115963511 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTGTGCTACTGAAATGTGGTGATG[C/T]AGCGAAGCCTGGTGAAAAAACAAAG
Phenotype
MIM: 608771
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MED13L PubMed Links

Gene Details

Gene
MED13L
Gene Name
mediator complex subunit 13 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015335.4 6451 Silent Mutation CTA,CTG L2132L NP_056150.1
XM_011538080.2 6451 Silent Mutation CTA,CTG L2144L XP_011536382.1
XM_011538081.2 6451 Silent Mutation CTA,CTG L2143L XP_011536383.1
XM_011538082.2 6451 Silent Mutation CTA,CTG L2134L XP_011536384.1
XM_017019090.1 6451 Silent Mutation CTA,CTG L2131L XP_016874579.1

View Full Product Details