Product Details

SNP ID
rs111810938
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:87675470 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAGCTGGATGCTGCAGACTCTAGCG[C/T]TTGCTGTAACATCTCTCGTCCTTTC
Phenotype
MIM: 609680
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MIR4500HG PubMed Links

Gene Details

Gene
MIR4500HG
Gene Name
MIR4500 host gene
There are no transcripts associated with this gene.

Gene
SLITRK5
Gene Name
SLIT and NTRK like family member 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015567.1 972 Missense Mutation CTT,TTT L28F NP_056382.1
XM_005254038.4 972 Missense Mutation CTT,TTT L28F XP_005254095.1
XM_005254039.2 972 Missense Mutation CTT,TTT L28F XP_005254096.1

View Full Product Details