Product Details

SNP ID
rs112107109
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:91350002 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTGGAAACTGGCTTATGCAGTTTA[C/T]GAAATATAATTTTAGACCCTCTTTT
Phenotype
MIM: 609416 MIM: 609415 MIM: 609417 MIM: 609418 MIM: 609419 MIM: 609420 MIM: 609422
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MIR17 PubMed Links

Gene Details

Gene
MIR17
Gene Name
microRNA 17
There are no transcripts associated with this gene.

Gene
MIR17HG
Gene Name
miR-17-92a-1 cluster host gene
There are no transcripts associated with this gene.

Gene
MIR18A
Gene Name
microRNA 18a
There are no transcripts associated with this gene.

Gene
MIR19A
Gene Name
microRNA 19a
There are no transcripts associated with this gene.

Gene
MIR19B1
Gene Name
microRNA 19b-1
There are no transcripts associated with this gene.

Gene
MIR20A
Gene Name
microRNA 20a
There are no transcripts associated with this gene.

Gene
MIR92A1
Gene Name
microRNA 92a-1
There are no transcripts associated with this gene.

View Full Product Details