Product Details

SNP ID
rs113261549
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:2818692 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTCTGCCAGCTATAGTGACCTTAG[C/T]GATCCCTCCGGGTGGATGGTCCAGT
Phenotype
MIM: 608159
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PRSS21 PubMed Links

Gene Details

Gene
PRSS21
Gene Name
protease, serine 21
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001270452.1 299 Silent Mutation AGC,AGT S89S NP_001257381.1
NM_006799.3 299 Silent Mutation AGC,AGT S91S NP_006790.1
NM_144956.2 299 Silent Mutation AGC,AGT S89S NP_659205.1
NM_144957.2 299 Silent Mutation AGC,AGT S91S NP_659206.1
XM_017022875.1 299 Silent Mutation AGC,AGT S89S XP_016878364.1
XM_017022876.1 299 Silent Mutation AGC,AGT S91S XP_016878365.1
XM_017022877.1 299 Silent Mutation AGC,AGT S89S XP_016878366.1
XM_017022878.1 299 Silent Mutation AGC,AGT S91S XP_016878367.1

View Full Product Details