Product Details

SNP ID
rs113845299
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:28477417 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTCCCAGACCTGCCGGGAAGGCTG[A/G]GAGCACAGTTCATGGAGGGTCTCTG
Phenotype
MIM: 607042
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CLN3 PubMed Links

Gene Details

Gene
CLN3
Gene Name
CLN3, battenin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000086.2 1775 UTR 3 NP_000077.1
NM_001042432.1 1775 UTR 3 NP_001035897.1
NM_001286104.1 1775 UTR 3 NP_001273033.1
NM_001286105.1 1775 UTR 3 NP_001273034.1
NM_001286109.1 1775 Intron NP_001273038.1
NM_001286110.1 1775 UTR 3 NP_001273039.1

View Full Product Details