Product Details
- SNP ID
-
rs111370319
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.16:3224016 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGGTCTCTCAGCCTCATGACTCTGC[A/G]GATGAAGCACAAACTCCTCATTCTT
- Phenotype
-
MIM: 603231
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
OR1F2P
PubMed Links
Gene Details
- Gene
- OR1F2P
- Gene Name
- olfactory receptor family 1 subfamily F member 2 pseudogene
There are no transcripts associated with this gene.
- Gene
- ZNF200
- Gene Name
- zinc finger protein 200
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001145446.1 |
1488 |
Missense Mutation |
CCG,CTG |
P354L |
NP_001138918.1 |
NM_001145447.1 |
1488 |
Missense Mutation |
CCG,CTG |
P354L |
NP_001138919.1 |
NM_001145448.1 |
1488 |
Missense Mutation |
CCG,CTG |
P354L |
NP_001138920.1 |
NM_003454.3 |
1488 |
Missense Mutation |
CCG,CTG |
P355L |
NP_003445.2 |
NM_198087.2 |
1488 |
Missense Mutation |
CCG,CTG |
P354L |
NP_932353.1 |
NM_198088.2 |
1488 |
Missense Mutation |
CCG,CTG |
P355L |
NP_932354.1 |
XM_005255556.1 |
1488 |
Missense Mutation |
CCG,CTG |
P355L |
XP_005255613.1 |
XM_006720940.2 |
1488 |
Missense Mutation |
CCG,CTG |
P355L |
XP_006721003.1 |
XM_006720941.2 |
1488 |
Missense Mutation |
CCG,CTG |
P355L |
XP_006721004.1 |
XM_017023647.1 |
1488 |
Missense Mutation |
CCG,CTG |
P355L |
XP_016879136.1 |
XM_017023648.1 |
1488 |
Intron |
|
|
XP_016879137.1 |
View Full Product Details