Product Details

SNP ID
rs117876257
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:593012 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAGAGGCAGAGAGGAGAGTCCACT[C/T]TTAGAAAGACACCACGTGCTCTCAG
Phenotype
MIM: 605754
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC101929280 PubMed Links

Gene Details

Gene
LOC101929280
Gene Name
uncharacterized LOC101929280
There are no transcripts associated with this gene.

Gene
PIGQ
Gene Name
phosphatidylinositol glycan anchor biosynthesis class Q
There are no transcripts associated with this gene.

Gene
RAB40C
Gene Name
RAB40C, member RAS oncogene family
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001172663.1 Intron NP_001166134.1
NM_001172664.1 Intron NP_001166135.1
NM_001172665.1 Intron NP_001166136.1
NM_001172666.1 Intron NP_001166137.1
NM_021168.4 Intron NP_066991.3

View Full Product Details