Product Details

SNP ID
rs116632380
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:81014228 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTATACTTAACCAATCAGTTTATTA[C/G]AGGCAATTTTGTTTCTTTCTTTGTG
Phenotype
MIM: 611509
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CENPN PubMed Links

Gene Details

Gene
CENPN
Gene Name
centromere protein N
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001100624.2 Intron NP_001094094.2
NM_001100625.2 Intron NP_001094095.2
NM_001270473.1 Intron NP_001257402.1
NM_001270474.1 Intron NP_001257403.1
NM_018455.5 Intron NP_060925.2
XM_006721236.3 Intron XP_006721299.1
XM_017023456.1 Intron XP_016878945.1
Gene
CMC2
Gene Name
C-X9-C motif containing 2
There are no transcripts associated with this gene.

View Full Product Details