Product Details

SNP ID
rs111830706
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.16:86510917 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCTACCCAAGGGCCTTGGGCGGCC[C/T]GGCAAGGGCCACTACTGGACCATCG
Phenotype
MIM: 614975 MIM: 601089
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
FENDRR PubMed Links

Gene Details

Gene
FENDRR
Gene Name
FOXF1 adjacent non-coding developmental regulatory RNA
There are no transcripts associated with this gene.

Gene
FOXF1
Gene Name
forkhead box F1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001451.2 391 Silent Mutation CCC,CCT P116P NP_001442.2

View Full Product Details