Product Details

SNP ID
rs117905209
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:52438211 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AACAGCATAAAAACACAATAAGCAA[C/T]GTAGCAATGAATGGTTTATGTCACC
Phenotype
MIM: 611416
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TOX3 PubMed Links

Gene Details

Gene
TOX3
Gene Name
TOX high mobility group box family member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001080430.3 2992 UTR 3 NP_001073899.2
NM_001146188.2 2992 UTR 3 NP_001139660.1
XM_005255892.3 2992 UTR 3 XP_005255949.1
XM_011523002.2 2992 UTR 3 XP_011521304.1
XM_017023142.1 2992 UTR 3 XP_016878631.1

View Full Product Details