Product Details

SNP ID
rs111931792
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.16:55846804 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAGGTAGCCCAGTATTTCATCATC[C/T]TCCGGCTCAGTAACTTCTCCTCCTC
Phenotype
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
CES5A PubMed Links

Gene Details

Gene
CES5A
Gene Name
carboxylesterase 5A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001143685.1 1609 Missense Mutation AAG,AGG K487R NP_001137157.1
NM_001190158.1 1609 Missense Mutation AAG,AGG K516R NP_001177087.1
NM_145024.2 1609 Missense Mutation AAG,AGG K437R NP_659461.1

View Full Product Details