Product Details

SNP ID
rs113937803
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:88428755 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGGCCTCCGGACACCGAGCTGGCC[A/G]CCCCAGGGCCCCCACCCGCCAGGCT
Phenotype
MIM: 612078
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF469 PubMed Links

Gene Details

Gene
ZNF469
Gene Name
zinc finger protein 469
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001127464.2 1478 Missense Mutation ACC,GCC T429A NP_001120936.2
XM_017023784.1 1478 Missense Mutation ACC,GCC T429A XP_016879273.1
XM_017023785.1 1478 Missense Mutation ACC,GCC T429A XP_016879274.1

View Full Product Details