Product Details

SNP ID
rs117555121
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:88428941 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCTTGGCCCCAAGTGCTCCCGACC[A/G]CCCGGCCAAGTCCCCACGGAATGGA
Phenotype
MIM: 612078
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF469 PubMed Links

Gene Details

Gene
ZNF469
Gene Name
zinc finger protein 469
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001127464.2 1664 Missense Mutation ACC,GCC T491A NP_001120936.2
XM_017023784.1 1664 Missense Mutation ACC,GCC T491A XP_016879273.1
XM_017023785.1 1664 Missense Mutation ACC,GCC T491A XP_016879274.1

View Full Product Details