Product Details

SNP ID
rs117065162
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:89162575 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGCGGCTGCACCGTGCTGTGGTGA[C/T]AGCCACAGCAGGATCAGAGCAGCCT
Phenotype
MIM: 614245 MIM: 114019
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ACSF3 PubMed Links

Gene Details

Gene
ACSF3
Gene Name
acyl-CoA synthetase family member 3
There are no transcripts associated with this gene.

Gene
CDH15
Gene Name
cadherin 15
There are no transcripts associated with this gene.

Gene
LINC00304
Gene Name
long intergenic non-protein coding RNA 304
There are no transcripts associated with this gene.

Gene
LOC400558
Gene Name
uncharacterized LOC400558
There are no transcripts associated with this gene.

View Full Product Details