Product Details

SNP ID
rs114082934
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:10681101 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCTCTTAGCAAGAGAATACTGCAT[C/T]CAGCAACACTGCTTTGGCTAGCTCT
Phenotype
MIM: 160720 MIM: 603644
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MYH3 PubMed Links

Gene Details

Gene
MYH3
Gene Name
myosin, heavy chain 3, skeletal muscle, embryonic
There are no transcripts associated with this gene.

Gene
SCO1
Gene Name
SCO1 cytochrome c oxidase assembly protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004589.3 985 UTR 3 NP_004580.1
XM_005256751.3 985 UTR 3 XP_005256808.1

View Full Product Details