Product Details

SNP ID
rs115407713
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:15031605 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTTGAGAGAGACAGACAGGGAGAG[A/G]GCATTGCTAGACATTCATCTGTACA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CDRT7 PubMed Links

Gene Details

Gene
CDRT7
Gene Name
CMT1A duplicated region transcript 7 (non-protein coding)
There are no transcripts associated with this gene.

View Full Product Details