Product Details

SNP ID
rs116194142
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:3514609 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGACAGTACCTGTTCGTCTTACAGG[A/C]CCCGGGTCTTCGTTAAGGAAGGAGA
Phenotype
MIM: 607066
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
SPATA22 PubMed Links

Gene Details

Gene
SPATA22
Gene Name
spermatogenesis associated 22
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001170695.1 2584 Intron NP_001164166.1
NM_001170696.1 2584 Intron NP_001164167.1
NM_001170697.1 2584 Intron NP_001164168.1
NM_001170698.1 2584 Intron NP_001164169.1
NM_001170699.1 2584 Intron NP_001164170.1
NM_001321336.1 2584 Intron NP_001308265.1
NM_001321337.1 2584 Intron NP_001308266.1
NM_032598.4 2584 Intron NP_115987.2
Gene
TRPV3
Gene Name
transient receptor potential cation channel subfamily V member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001258205.1 2584 Silent Mutation GGG,GGT G754G NP_001245134.1
NM_145068.3 2584 Silent Mutation GGG,GGT G754G NP_659505.1

View Full Product Details