Product Details

SNP ID
rs115900148
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:9772889 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCCACTTCGGCTCGGAGGCAGTCA[A/G]AGAAGCCCAGGGCTGCGTGCTTGGA
Phenotype
MIM: 616161
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DHRS7C PubMed Links

Gene Details

Gene
DHRS7C
Gene Name
dehydrogenase/reductase 7C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001105571.2 618 Missense Mutation TCT,TTT S202F NP_001099041.1
NM_001220493.1 618 Missense Mutation TCT,TTT S203F NP_001207422.1

View Full Product Details