Product Details

SNP ID
rs115882848
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:53823014 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCCCAAGGGCCCTGGAGCGCTCCC[G/T]GATACCTCCATCACTCACCATGGCC
Phenotype
MIM: 615142
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
KIF2B PubMed Links

Gene Details

Gene
KIF2B
Gene Name
kinesin family member 2B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032559.4 137 UTR 5 NP_115948.4

View Full Product Details