Product Details

SNP ID
rs115025206
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:39921724 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCTGTGGGTAAATTAACCCTCCAA[A/G]CCTGCTTCCCTCCCTGCCCACATCC
Phenotype
MIM: 611221 MIM: 610075
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GSDMB PubMed Links

Gene Details

Gene
GSDMB
Gene Name
gasdermin B
There are no transcripts associated with this gene.

Gene
LRRC3C
Gene Name
leucine rich repeat containing 3C
There are no transcripts associated with this gene.

Gene
ORMDL3
Gene Name
ORMDL sphingolipid biosynthesis regulator 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001320801.1 1323 UTR 3 NP_001307730.1
NM_001320802.1 1323 UTR 3 NP_001307731.1
NM_001320803.1 1323 UTR 3 NP_001307732.1
NM_139280.3 1323 UTR 3 NP_644809.1
XM_017025345.1 1323 UTR 3 XP_016880834.1
XM_017025346.1 1323 UTR 3 XP_016880835.1
XM_017025347.1 1323 UTR 3 XP_016880836.1
XM_017025348.1 1323 UTR 3 XP_016880837.1

View Full Product Details