Product Details

SNP ID
rs111474879
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:40364086 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATCCCCCATTTGCATTTTAGCCGCA[C/T]CCCCTGAGCCGTCTCCGTTCGACCC
Phenotype
MIM: 607425 MIM: 180240
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GJD3 PubMed Links

Gene Details

Gene
GJD3
Gene Name
gap junction protein delta 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152219.3 608 UTR 5 NP_689343.3
Gene
RARA
Gene Name
retinoic acid receptor alpha
There are no transcripts associated with this gene.

View Full Product Details