Product Details

SNP ID
rs114164544
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:40777313 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATTTAGAACAGGAGCTGTAAAAGT[A/G]TAGAGCGCTTATATTAATTATTCTG
Phenotype
MIM: 616675 MIM: 616676
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KRT26 PubMed Links

Gene Details

Gene
KRT26
Gene Name
keratin 26
There are no transcripts associated with this gene.

Gene
KRT27
Gene Name
keratin 27
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_181537.3 Intron NP_853515.2

View Full Product Details