Product Details

SNP ID
rs111862238
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:40977800 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTAAGATATATGACATTTCTCATGG[A/G]TTCATTAATTTATTTGTATTTGAGG
Phenotype
MIM: 616679
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KRT40 PubMed Links

Gene Details

Gene
KRT40
Gene Name
keratin 40
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_182497.3 1854 UTR 3 NP_872303.2
XM_011524339.2 1854 Intron XP_011522641.2
XM_017024189.1 1854 UTR 3 XP_016879678.1

View Full Product Details