Product Details

SNP ID
rs117840247
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:81516180 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATCTTCCACCACTAAATGAAACGAT[A/G]GACCCTGAACAGAAAAAGGAACAGT
Phenotype
MIM: 102560 MIM: 607643
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ACTG1 PubMed Links

Gene Details

Gene
ACTG1
Gene Name
actin gamma 1
There are no transcripts associated with this gene.

Gene
FSCN2
Gene Name
fascin actin-bundling protein 2, retinal
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001077182.2 2533 Intron NP_001070650.1
NM_012418.3 2533 Intron NP_036550.1
XM_011524587.2 2533 Intron XP_011522889.1
XM_011524590.2 2533 UTR 5 XP_011522892.1

View Full Product Details