Product Details

SNP ID
rs117218057
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:43175980 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAGGTAGCTATTTTTAGCAGTGTT[A/G]TTTCATAGCTGTGAAGTCTGTCATA
Phenotype
MIM: 166945
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LOC101929767 PubMed Links

Gene Details

Gene
LOC101929767
Gene Name
uncharacterized LOC101929767
There are no transcripts associated with this gene.

Gene
NBR1
Gene Name
NBR1, autophagy cargo receptor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001291571.1 Intron NP_001278500.1
NM_001291572.1 Intron NP_001278501.1
NM_005899.4 Intron NP_005890.2
NM_031862.3 Intron NP_114068.1
XM_006721903.1 Intron XP_006721966.1
XM_011524813.1 Intron XP_011523115.1
XM_017024642.1 Intron XP_016880131.1
XM_017024643.1 Intron XP_016880132.1
XM_017024644.1 Intron XP_016880133.1

View Full Product Details