Product Details

SNP ID
rs117569358
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:44852701 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAGAGTAACCAAGAATGTTCTACA[A/G]AAAGCAGAAAAGGCTCAGGGCTCTC
Phenotype
MIM: 603892
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
EFTUD2 PubMed Links

Gene Details

Gene
EFTUD2
Gene Name
elongation factor Tu GTP binding domain containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001142605.1 Intron NP_001136077.1
NM_001258353.1 Intron NP_001245282.1
NM_001258354.1 Intron NP_001245283.1
NM_004247.3 Intron NP_004238.3
Gene
HIGD1B
Gene Name
HIG1 hypoxia inducible domain family member 1B
There are no transcripts associated with this gene.

View Full Product Details