Product Details

SNP ID
rs113215819
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:45164848 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATGAGCAGGGGGCTTAGGACAGAG[A/G]TCAGAGAGCTTCTGACCCACCCTGC
Phenotype
MIM: 615695
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
HEXIM2 PubMed Links

Gene Details

Gene
HEXIM2
Gene Name
hexamethylene bisacetamide inducible 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001303436.1 Intron NP_001290365.1
NM_001303437.1 Intron NP_001290366.1
NM_001303438.1 Intron NP_001290367.1
NM_001303439.1 Intron NP_001290368.1
NM_001303440.1 Intron NP_001290369.1
NM_001303441.1 Intron NP_001290370.1
NM_001303442.1 Intron NP_001290371.1
NM_001303443.1 Intron NP_001290372.1
NM_001303444.1 Intron NP_001290373.1
NM_144608.2 Intron NP_653209.1
XM_006721687.3 Intron XP_006721750.1
XM_011524306.2 Intron XP_011522608.1
XM_011524307.2 Intron XP_011522609.2
XM_011524308.2 Intron XP_011522610.2
XM_017024167.1 Intron XP_016879656.1
XM_017024168.1 Intron XP_016879657.1
XM_017024169.1 Intron XP_016879658.1
XM_017024170.1 Intron XP_016879659.1
Gene
LOC105371795
Gene Name
uncharacterized LOC105371795
There are no transcripts associated with this gene.

View Full Product Details