Product Details
- SNP ID
-
rs112826052
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:45169130 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TCAGAGGATGAAGATCTTGCTGGGG[C/T]TGTCGGTGGCCTGGGCTGGAACAGT
- Phenotype
-
MIM: 615695
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
HEXIM2
PubMed Links
Gene Details
- Gene
- HEXIM2
- Gene Name
- hexamethylene bisacetamide inducible 2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001303436.1 |
1301 |
Missense Mutation |
GCT,GTT |
A83V |
NP_001290365.1 |
NM_001303437.1 |
1301 |
Missense Mutation |
GCT,GTT |
A61V |
NP_001290366.1 |
NM_001303438.1 |
1301 |
Missense Mutation |
GCT,GTT |
A61V |
NP_001290367.1 |
NM_001303439.1 |
1301 |
Missense Mutation |
GCT,GTT |
A61V |
NP_001290368.1 |
NM_001303440.1 |
1301 |
Missense Mutation |
GCT,GTT |
A61V |
NP_001290369.1 |
NM_001303441.1 |
1301 |
Missense Mutation |
GCT,GTT |
A61V |
NP_001290370.1 |
NM_001303442.1 |
1301 |
Missense Mutation |
GCT,GTT |
A61V |
NP_001290371.1 |
NM_001303443.1 |
1301 |
Missense Mutation |
GCT,GTT |
A61V |
NP_001290372.1 |
NM_001303444.1 |
1301 |
Missense Mutation |
GCT,GTT |
A61V |
NP_001290373.1 |
NM_144608.2 |
1301 |
Missense Mutation |
GCT,GTT |
A61V |
NP_653209.1 |
XM_006721687.3 |
1301 |
Missense Mutation |
GCT,GTT |
A84V |
XP_006721750.1 |
XM_011524306.2 |
1301 |
Missense Mutation |
GCT,GTT |
A61V |
XP_011522608.1 |
XM_011524307.2 |
1301 |
Missense Mutation |
GCT,GTT |
A221V |
XP_011522609.2 |
XM_011524308.2 |
1301 |
Missense Mutation |
GCT,GTT |
A222V |
XP_011522610.2 |
XM_017024167.1 |
1301 |
Missense Mutation |
GCT,GTT |
A61V |
XP_016879656.1 |
XM_017024168.1 |
1301 |
Missense Mutation |
GCT,GTT |
A61V |
XP_016879657.1 |
XM_017024169.1 |
1301 |
Missense Mutation |
GCT,GTT |
A61V |
XP_016879658.1 |
XM_017024170.1 |
1301 |
Missense Mutation |
GCT,GTT |
A61V |
XP_016879659.1 |
- Gene
- LOC105371795
- Gene Name
- uncharacterized LOC105371795
There are no transcripts associated with this gene.
View Full Product Details