Product Details

SNP ID
rs111905881
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:31852274 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGCACAGCACACCCAAGGTTTAAG[C/T]TCTTGAGAAATGCTCTCCTGGATGT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
COPRS PubMed Links

Gene Details

Gene
COPRS
Gene Name
coordinator of PRMT5 and differentiation stimulator
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018405.3 528 Silent Mutation GAA,GAG E140E NP_060875.2
XM_005258000.1 528 Silent Mutation GAA,GAG E128E XP_005258057.1

View Full Product Details