Product Details

SNP ID
rs114851130
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:31852295 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAAGCTCTTGAGAAATGCTCTCCTG[A/G]ATGTCGTCAGCATCTGCAGGCAGTT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
COPRS PubMed Links

Gene Details

Gene
COPRS
Gene Name
coordinator of PRMT5 and differentiation stimulator
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018405.3 507 Silent Mutation ATC,ATT I133I NP_060875.2
XM_005258000.1 507 Silent Mutation ATC,ATT I121I XP_005258057.1

View Full Product Details