Product Details

SNP ID
rs117928919
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:11884215 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATGCAACCTTTGATGATACATATA[A/T]TTGATAAAAATGAGAAAACAGATTT
Phenotype
MIM: 139312 MIM: 611900
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
GNAL PubMed Links

Gene Details

Gene
GNAL
Gene Name
G protein subunit alpha L
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001142339.2 2708 UTR 3 NP_001135811.1
NM_001261443.1 2708 UTR 3 NP_001248372.1
NM_001261444.1 2708 UTR 3 NP_001248373.1
NM_182978.3 2708 UTR 3 NP_892023.1
XM_006722324.2 2708 Intron XP_006722387.1
Gene
MPPE1
Gene Name
metallophosphoesterase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001242904.1 2708 UTR 3 NP_001229833.1
NM_001319154.1 2708 UTR 3 NP_001306083.1
NM_023075.5 2708 UTR 3 NP_075563.3
XM_006722340.3 2708 UTR 3 XP_006722403.1
XM_006722341.3 2708 UTR 3 XP_006722404.1
XM_006722342.3 2708 UTR 3 XP_006722405.1
XM_006722343.3 2708 UTR 3 XP_006722406.1
XM_006722345.3 2708 UTR 3 XP_006722408.1
XM_006722346.2 2708 UTR 3 XP_006722409.1
XM_006722347.3 2708 UTR 3 XP_006722410.1
XM_006722348.2 2708 Intron XP_006722411.1
XM_011525731.2 2708 UTR 3 XP_011524033.1
XM_011525732.1 2708 UTR 3 XP_011524034.1
XM_011525734.1 2708 UTR 3 XP_011524036.1
XM_017025920.1 2708 UTR 3 XP_016881409.1
XM_017025921.1 2708 UTR 3 XP_016881410.1
XM_017025922.1 2708 UTR 3 XP_016881411.1
XM_017025923.1 2708 UTR 3 XP_016881412.1
XM_017025924.1 2708 UTR 3 XP_016881413.1
XM_017025925.1 2708 UTR 3 XP_016881414.1
XM_017025926.1 2708 UTR 3 XP_016881415.1
XM_017025927.1 2708 UTR 3 XP_016881416.1
XM_017025928.1 2708 UTR 3 XP_016881417.1
XM_017025929.1 2708 UTR 3 XP_016881418.1
XM_017025930.1 2708 UTR 3 XP_016881419.1
XM_017025931.1 2708 Intron XP_016881420.1

View Full Product Details