Product Details

SNP ID
rs117052266
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:35367595 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTTTTATCAAGAATCTCTGGAAAAT[A/G]GATAATTTTGTTGTTTGTATACTGT
Phenotype
MIM: 609600
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF396 PubMed Links

Gene Details

Gene
ZNF396
Gene Name
zinc finger protein 396
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001322286.1 1890 UTR 3 NP_001309215.1
NM_001322290.1 1890 UTR 3 NP_001309219.1
NM_001322291.1 1890 Intron NP_001309220.1
NM_145756.2 1890 UTR 3 NP_665699.1
XM_006722432.3 1890 Intron XP_006722495.1
XM_017025687.1 1890 Intron XP_016881176.1
XM_017025688.1 1890 Intron XP_016881177.1
XM_017025689.1 1890 Intron XP_016881178.1

View Full Product Details