Product Details
- SNP ID
-
rs114831324
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.18:54360683 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GTAATTCCTCTAATTCTGATGGCTC[A/G]TTTCACTATAAAGATAAGCTGTACA
- Phenotype
-
MIM: 613258
MIM: 607051
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
C18orf54
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs1657904] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- C18orf54
- Gene Name
- chromosome 18 open reading frame 54
- Gene
- STARD6
- Gene Name
- StAR related lipid transfer domain containing 6
There are no transcripts associated with this gene.
View Full Product Details