Product Details
- SNP ID
-
rs117975009
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:37337152 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGAACAACGTCTTTATCATTTGGAC[A/G]TGACCTTCAGAGTATCAGCTGCACT
- Phenotype
-
MIM: 165250
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
HKR1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs144190488] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- HKR1
- Gene Name
- HKR1, GLI-Kruppel zinc finger family member
- Gene
- LOC105372392
- Gene Name
- uncharacterized LOC105372392
There are no transcripts associated with this gene.
View Full Product Details