Product Details

SNP ID
rs112385179
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:39480889 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGGCAGCGGTGTTCTCGCGCTTGCG[A/G]AGCGGGCTCCGGCTCGGCTCGCGGG
Phenotype
MIM: 602102 MIM: 607381
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SUPT5H PubMed Links

Gene Details

Gene
SUPT5H
Gene Name
SPT5 homolog, DSIF elongation factor subunit
There are no transcripts associated with this gene.

Gene
TIMM50
Gene Name
translocase of inner mitochondrial membrane 50
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_011527491.2 44 Intron XP_011525793.1
XM_017027472.1 44 UTR 5 XP_016882961.1

View Full Product Details