Product Details

SNP ID
rs117361485
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:39731420 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTGACATTAAATTTGGTCAGGGAG[A/C]TATCTCTCCACACTTGCACCATCTT
Phenotype
MIM: 153310
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
CLC PubMed Links

Gene Details

Gene
CLC
Gene Name
Charcot-Leyden crystal galectin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001828.5 Intron NP_001819.2

View Full Product Details