Product Details

SNP ID
rs114240382
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:43597167 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGCAGGATTCACCCAAGGAGAGAA[A/G]TCCCCAGAGCCCAGGAGGCAACATC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
IRGQ PubMed Links

Gene Details

Gene
IRGQ
Gene Name
immunity related GTPase Q
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001007561.2 232 Intron NP_001007562.1
XM_005258515.3 232 Intron XP_005258572.1
Gene
ZNF576
Gene Name
zinc finger protein 576
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145347.1 232 Missense Mutation AAT,AGT N20S NP_001138819.1
NM_024327.2 232 Missense Mutation AAT,AGT N20S NP_077303.1

View Full Product Details