Product Details

SNP ID
rs114151928
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:44699739 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAACAGGCATACCAATCAGACCCAC[A/G]TCCACCAGGCTCTGGCTTAAATCCC
Phenotype
MIM: 614591
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CEACAM16 PubMed Links

Gene Details

Gene
CEACAM16
Gene Name
carcinoembryonic antigen related cell adhesion molecule 16
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001039213.3 Intron NP_001034302.2
XM_017026795.1 Intron XP_016882284.1

View Full Product Details