Product Details

SNP ID
rs117289933
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:45407107 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTCTGCTGCAGATGCTGCTCGGTT[C/G]TCTTGTCCCCCCAACTTTACCGCGA
Phenotype
MIM: 107325 MIM: 126380 MIM: 607463
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CD3EAP PubMed Links

Gene Details

Gene
CD3EAP
Gene Name
CD3e molecule associated protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001297590.1 610 Missense Mutation TTC,TTG F14L NP_001284519.1
NM_012099.1 610 Missense Mutation TTC,TTG F12L NP_036231.1
XM_017026180.1 610 UTR 5 XP_016881669.1
XM_017026181.1 610 UTR 5 XP_016881670.1
Gene
ERCC1
Gene Name
ERCC excision repair 1, endonuclease non-catalytic subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001166049.1 610 Intron NP_001159521.1
NM_001983.3 610 Intron NP_001974.1
NM_202001.2 610 Intron NP_973730.1
XM_005258634.1 610 Intron XP_005258691.1
XM_005258635.2 610 Intron XP_005258692.1
XM_005258636.4 610 Intron XP_005258693.1
XM_005258637.1 610 Intron XP_005258694.1
XM_011526610.2 610 Intron XP_011524912.1
XM_017026459.1 610 Intron XP_016881948.1
XM_017026460.1 610 Intron XP_016881949.1
XM_017026461.1 610 Intron XP_016881950.1
XM_017026462.1 610 Intron XP_016881951.1
XM_017026463.1 610 Intron XP_016881952.1
XM_017026464.1 610 Intron XP_016881953.1
XM_017026465.1 610 Intron XP_016881954.1
XM_017026466.1 610 Intron XP_016881955.1
Gene
PPP1R13L
Gene Name
protein phosphatase 1 regulatory subunit 13 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001142502.1 610 Intron NP_001135974.1
NM_006663.3 610 Intron NP_006654.2
XM_017026177.1 610 Intron XP_016881666.1
XM_017026178.1 610 Intron XP_016881667.1
XM_017026179.1 610 Intron XP_016881668.1

View Full Product Details