Product Details
- SNP ID
-
rs116223719
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:49364655 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TCCTCTCTACCCTGGTGATCCCCTC[C/T]GCTGCAGCTCCTATCCATGATGCTG
- Phenotype
-
MIM: 605418
MIM: 601729
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
DKKL1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs919364] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- DKKL1
- Gene Name
- dickkopf like acrosomal protein 1
- Gene
- LOC101928295
- Gene Name
- uncharacterized LOC101928295
There are no transcripts associated with this gene.
- Gene
- TEAD2
- Gene Name
- TEA domain transcription factor 2
There are no transcripts associated with this gene.
View Full Product Details