Product Details

SNP ID
rs112077618
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:49423273 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCAGTGCGGACGCCCCAGGGCACCA[C/G]CAGCAGCAGCAGCAGCAGCAGCAGC
Phenotype
MIM: 608386 MIM: 605208
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CCDC155 PubMed Links

Gene Details

Gene
CCDC155
Gene Name
coiled-coil domain containing 155
There are no transcripts associated with this gene.

Gene
GFY
Gene Name
golgi associated olfactory signaling regulator
There are no transcripts associated with this gene.

Gene
PTH2
Gene Name
parathyroid hormone 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_178449.3 169 Missense Mutation CTG,GTG L23V NP_848544.1
Gene
SLC17A7
Gene Name
solute carrier family 17 member 7
There are no transcripts associated with this gene.

View Full Product Details