Product Details
- SNP ID
-
rs115151289
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:51965074 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGCCACAGAAGGCACTTGTGTAGTC[A/G]CCCCGTTAGCAGAGTTTTTCTCCTG
- Phenotype
-
MIM: 605422
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ZNF350
PubMed Links
Gene Details
- Gene
- ZNF350
- Gene Name
- zinc finger protein 350
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_021632.3 |
1766 |
Missense Mutation |
GCG,GTG |
A460V |
NP_067645.3 |
XM_011527196.1 |
1766 |
Missense Mutation |
GCG,GTG |
A424V |
XP_011525498.1 |
XM_017027094.1 |
1766 |
Missense Mutation |
GCG,GTG |
A502V |
XP_016882583.1 |
XM_017027095.1 |
1766 |
Missense Mutation |
GCG,GTG |
A502V |
XP_016882584.1 |
XM_017027096.1 |
1766 |
Missense Mutation |
GCG,GTG |
A502V |
XP_016882585.1 |
XM_017027097.1 |
1766 |
Missense Mutation |
GCG,GTG |
A466V |
XP_016882586.1 |
XM_017027098.1 |
1766 |
Missense Mutation |
GCG,GTG |
A460V |
XP_016882587.1 |
XM_017027099.1 |
1766 |
Missense Mutation |
GCG,GTG |
A460V |
XP_016882588.1 |
XM_017027100.1 |
1766 |
Missense Mutation |
GCG,GTG |
A460V |
XP_016882589.1 |
XM_017027101.1 |
1766 |
Missense Mutation |
GCG,GTG |
A234V |
XP_016882590.1 |
XM_017027102.1 |
1766 |
Missense Mutation |
GCG,GTG |
A234V |
XP_016882591.1 |
XM_017027103.1 |
1766 |
Missense Mutation |
GCG,GTG |
A234V |
XP_016882592.1 |
XM_017027104.1 |
1766 |
Missense Mutation |
GCG,GTG |
A234V |
XP_016882593.1 |
- Gene
- ZNF350-AS1
- Gene Name
- ZNF350 antisense RNA 1
There are no transcripts associated with this gene.
View Full Product Details