Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001102603.1 | 4723 | Intron | NP_001096073.1 | ||
NM_001322125.1 | 4723 | Intron | NP_001309054.1 | ||
NM_001322126.1 | 4723 | Intron | NP_001309055.1 | ||
NM_001322128.1 | 4723 | Intron | NP_001309057.1 | ||
NM_001322129.1 | 4723 | Intron | NP_001309058.1 | ||
NM_001322130.1 | 4723 | Intron | NP_001309059.1 | ||
NM_001322131.1 | 4723 | Intron | NP_001309060.1 | ||
NM_001322132.1 | 4723 | Intron | NP_001309061.1 | ||
NM_001322133.1 | 4723 | Intron | NP_001309062.1 | ||
NM_001322134.1 | 4723 | Intron | NP_001309063.1 | ||
NM_001322135.1 | 4723 | Intron | NP_001309064.1 | ||
NM_001322136.1 | 4723 | Intron | NP_001309065.1 | ||
NM_001322137.1 | 4723 | Intron | NP_001309066.1 | ||
NM_001322138.1 | 4723 | Intron | NP_001309067.1 | ||
NM_001322139.1 | 4723 | Intron | NP_001309068.1 | ||
NM_033288.3 | 4723 | Intron | NP_150630.2 | ||
NM_198893.2 | 4723 | Intron | NP_942596.1 | ||
XM_017027445.1 | 4723 | UTR 3 | XP_016882934.1 | ||
XM_017027446.1 | 4723 | UTR 3 | XP_016882935.1 | ||
XM_017027447.1 | 4723 | Intron | XP_016882936.1 | ||
XM_017027448.1 | 4723 | Intron | XP_016882937.1 |