Product Details

SNP ID
rs114465612
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:55649275 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACCCCACCTAGACCCACAGATCTT[C/T]TACAGTCTGAGCCCCTCTCGGAGAA
Phenotype
MIM: 613478 MIM: 191318
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CCDC106 PubMed Links

Gene Details

Gene
CCDC106
Gene Name
coiled-coil domain containing 106
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_013301.2 456 Silent Mutation TTC,TTT F34F NP_037433.2
XM_005258827.2 456 Silent Mutation TTC,TTT F34F XP_005258884.1
XM_005258828.2 456 Silent Mutation TTC,TTT F34F XP_005258885.1
Gene
LOC107983998
Gene Name
uncharacterized LOC107983998
There are no transcripts associated with this gene.

Gene
U2AF2
Gene Name
U2 small nuclear RNA auxiliary factor 2
There are no transcripts associated with this gene.

Gene
ZNF580
Gene Name
zinc finger protein 580
There are no transcripts associated with this gene.

Gene
ZNF581
Gene Name
zinc finger protein 581
There are no transcripts associated with this gene.

View Full Product Details