Product Details
- SNP ID
-
rs114188545
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:30224761 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TACGCCACAAGGTTACAGCGCGGAT[A/C]AACGTCATTTGATCGCTGATGACTC
- Phenotype
-
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
ZNF536
PubMed Links
Gene Details
- Gene
- ZNF536
- Gene Name
- zinc finger protein 536
View Full Product Details